siRNA Details
| Gene Symbol: Gm38572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054526.1 | - | 4 | 575 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ053110.1 | - | 3 | 332 | Chr. 8 - 31601135 - 31663828 [-] on Build GRCm38 | |

Gm38572
Genomic Map