siRNA Details
| Gene Symbol: 4921529L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110484.1 | 4 | 654 | |||
| GenBank mRNA | AK076597.1 | - | 4 | 654 | ||
| AK085171.1 | - | 487 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921529L05Rik| Gene Symbol: 4921529L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110484.1 | 4 | 654 | |||
| GenBank mRNA | AK076597.1 | - | 4 | 654 | ||
| AK085171.1 | - | 487 | ||||