siRNA Details
| Gene Symbol: 4921529L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110484.1 | 3 | 267 | |||
| GenBank mRNA | AK076597.1 | - | 3 | 267 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB614683.1 | - | 3 | 267 | Chr. 6 - 53978691 - 54016518 [+] on Build GRCm38 | |

4921529L05Rik
Genomic Map