siRNA Details
Gene Symbol: Gm33101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131736.1 | - | 1, 2 | 72 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706661.1 | - | 1, 2 | 77 | Chr. 5 - 106369012 - 106369718 [+] on Build GRCm38 |
