siRNA Details
| Gene Symbol: Gm33101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131736.1 | - | 2 | 360 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706661.1 | - | 2 | 365 | Chr. 5 - 106369012 - 106369718 [+] on Build GRCm38 | |

Gm33101
Genomic Map