siRNA Details
Gene Symbol: Gm33101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131736.1 | - | 3, 4 | 974 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV045176.2 | - | 2, 3 | 201 | Chr. 5 - 106369868 - 106370818 [+] on Build GRCm38 |
