siRNA Details
Gene Symbol: 4921507L20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045827.1 | 4 | 1324 | |||
GenBank mRNA | AK014837.1 | - | 4 | 1324 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4921507L20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045827.1 | 4 | 1324 | |||
GenBank mRNA | AK014837.1 | - | 4 | 1324 |