siRNA Details
| Gene Symbol: Gm553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC051519.1 | - | 245 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI121625.1 | - | 1 | 248 | Chr. 1 - 52522899 - 52582505 [+] on Build GRCm38 | |

Gm553
Genomic Map