siRNA Details
Gene Symbol: Gm553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC051519.1 | - | 197 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI121625.1 | - | 1 | 200 | Chr. 1 - 52522899 - 52582505 [+] on Build GRCm38 |
