siRNA Details
| Gene Symbol: Gm11651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104047.1 | 3 | 614 | |||
| GenBank mRNA | AK016887.1 | - | 3 | 614 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716566.1 | - | 3 | 614 | Chr. 11 - 105965903 - 105985694 [-] on Build GRCm38 | |

Gm11651
Genomic Map