siRNA Details
| Gene Symbol: 1700054A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045320.1 | 1 | 179 | |||
| GenBank mRNA | AK018858.1 | - | 1 | 180 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706762.1 | - | 1 | 180 | Chr. 19 - 53076252 - 53084266 [+] on Build GRCm38 | |

1700054A03Rik
Genomic Map