siRNA Details
| Gene Symbol: 1700057H15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040774.1 | 4 | 381 | |||
| GenBank mRNA | AK132134.1 | - | 4 | 381 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY713246.1 | - | 4 | 381 | Chr. 4 - 124449902 - 124485959 [-] on Build GRCm38 | |

1700057H15Rik
Genomic Map