siRNA Details
| Gene Symbol: Igkv9-129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY498738.1 | - | 1 | 226 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AB089682.1 | - | 226 | Chr. - - [-] on Build | ||

Igkv9-129
Genomic Map