siRNA Details
| Gene Symbol: Igkv9-129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY498738.1 | - | 1 | 267 | ||
| Gene Symbol: Igkv9-120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF124721.1 | - | 327 | |||
| U88675.1 | - | 369 | ||||

Igkv9-129
Genomic Map