siRNA Details
| Gene Symbol: 1700052M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018857.1 | - | 1 | 123 | ||
| AV046246.3 | - | 1 | 139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700052M18Rik| Gene Symbol: 1700052M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018857.1 | - | 1 | 123 | ||
| AV046246.3 | - | 1 | 139 | |||