siRNA Details
| Gene Symbol: 3110038A09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014137.1 | - | 995 | |||
| AV165602.1 | - | 1 | 272 | |||
| AV165624.1 | - | 1 | 277 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK014161.1 | - | 582 | Chr. - - [-] on Build | ||

3110038A09Rik
Genomic Map