siRNA Details
| Gene Symbol: 2610024L02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011538.1 | - | 1 | 85 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY710795.1 | - | 1 | 85 | Chr. 6 - 120835382 - 120836074 [-] on Build GRCm38 | |

2610024L02Rik
Genomic Map