siRNA Details
| Gene Symbol: 2610024J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011537.1 | - | 1 | 17 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY710793.1 | - | 1 | 17 | Chr. 4 - 6941878 - 6942298 [-] on Build GRCm38 | |

2610024J18Rik
Genomic Map