siRNA Details
| Gene Symbol: Gm2233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC035543.1 | - | 2 | 1279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2233| Gene Symbol: Gm2233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC035543.1 | - | 2 | 1279 | ||