siRNA Details
| Gene Symbol: Gm36017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076918.1 | - | 4 | 776 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB015460.2 | - | 3 | 469 | Chr. 5 - 44412793 - 44426992 [-] on Build GRCm38 | |

Gm36017
Genomic Map