siRNA Details
| Gene Symbol: Gm36017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076918.1 | - | 1 | 138 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615740.1 | - | 1 | 138 | Chr. 5 - 44413664 - 44427590 [-] on Build GRCm38 | |

Gm36017
Genomic Map