siRNA Details
Gene Symbol: Gm11269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132790.1 | - | 3 | 497 | ||
AV258989.2 | - | 1 | 91 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614867.1 | - | 3 | 494 | Chr. 4 - 82692805 - 82693711 [+] on Build GRCm38 |
