siRNA Details
| Gene Symbol: 4930597G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045736.1 | 4 | 476 | |||
| GenBank mRNA | AK016405.1 | - | 4 | 477 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716056.1 | - | 4 | 477 | Chr. 14 - 51317531 - 51345489 [-] on Build GRCm38 | |

4930597G03Rik
Genomic Map