siRNA Details
| Gene Symbol: 4930597G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045736.1 | 5 | 797 | |||
| GenBank mRNA | AK016405.1 | - | 5 | 798 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930597G03Rik| Gene Symbol: 4930597G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045736.1 | 5 | 797 | |||
| GenBank mRNA | AK016405.1 | - | 5 | 798 | ||