siRNA Details
| Gene Symbol: D16Ertd519e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040474.1 | 3 | 483 | |||
| GenBank mRNA | AK139678.1 | - | 3 | 484 | ||
| CO811502.1 | - | 3 | 492 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D16Ertd519e| Gene Symbol: D16Ertd519e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040474.1 | 3 | 483 | |||
| GenBank mRNA | AK139678.1 | - | 3 | 484 | ||
| CO811502.1 | - | 3 | 492 | |||