siRNA Details
| Gene Symbol: Gm11627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040286.1 | 3 | 321 | |||
| GenBank mRNA | AK144330.1 | - | 3 | 321 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11627| Gene Symbol: Gm11627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040286.1 | 3 | 321 | |||
| GenBank mRNA | AK144330.1 | - | 3 | 321 | ||