siRNA Details
| Gene Symbol: Gm11627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040286.1 | 1 | 51 | |||
| GenBank mRNA | AK144330.1 | - | 1 | 51 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB870579.1 | - | 1 | 51 | Chr. 11 - 102576714 - 102579119 [-] on Build GRCm38 | |

Gm11627
Genomic Map