siRNA Details
Gene Symbol: Gm11627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040286.1 | 3 | 350 | |||
GenBank mRNA | AK144330.1 | - | 3 | 350 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm11627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040286.1 | 3 | 350 | |||
GenBank mRNA | AK144330.1 | - | 3 | 350 |