siRNA Details
| Gene Symbol: 2900064K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013751.1 | - | 1 | 727 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712719.1 | - | 1 | 723 | Chr. 5 - 49518002 - 49518933 [-] on Build GRCm38 | |

2900064K03Rik
Genomic Map