siRNA Details
| Gene Symbol: 4930540M03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040746.1 | 5 | 361 | |||
| GenBank mRNA | AK019729.1 | - | 5 | 363 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715591.1 | - | 5 | 363 | Chr. 9 - 15619857 - 15641224 [+] on Build GRCm38 | |

4930540M03Rik
Genomic Map