siRNA Details
| Gene Symbol: 4930538L07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019723.1 | - | 1 | 152 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715567.1 | - | 1 | 152 | Chr. 14 - 47757034 - 47757498 [-] on Build GRCm38 | |

4930538L07Rik
Genomic Map