siRNA Details
| Gene Symbol: 4930538E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019722.1 | - | 3 | 432 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715563.1 | - | 3 | 432 | Chr. 11 - 24166873 - 24172781 [-] on Build GRCm38 | |

4930538E20Rik
Genomic Map