siRNA Details
| Gene Symbol: Ppp1r9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013208.1 | - | 1 | 210 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r9a| Gene Symbol: Ppp1r9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013208.1 | - | 1 | 210 | ||