siRNA Details
| Gene Symbol: 4930539C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019724.1 | - | 1 | 348 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715570.1 | - | 1 | 348 | Chr. 9 - 23749902 - 23750322 [+] on Build GRCm38 | |

4930539C01Rik
Genomic Map