siRNA Details
| Gene Symbol: Gm29999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC100375.1 | - | 1 | 32 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CO800717.1 | - | 1 | 35 | Chr. 3 - 82660539 - 82664902 [-] on Build GRCm38 | |

Gm29999
Genomic Map