siRNA Details
| Gene Symbol: Gm29999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC100375.1 | - | 2 | 648 | ||
| Gene Symbol: AA419673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136179.1 | - | 2713 | |||

Gm29999
Genomic Map