siRNA Details
| Gene Symbol: 1700026F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045487.1 | 2 | 269 | |||
| GenBank mRNA | AK006380.1 | - | 2 | 270 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706443.1 | - | 2 | 267 | Chr. 8 - 71006728 - 71026753 [-] on Build GRCm38 | |

1700026F02Rik
Genomic Map