siRNA Details
| Gene Symbol: Ncam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053403.1 | - | 1 | 2677 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB538918.2 | - | 1 | 364 | Chr. 9 - 49795893 - 49796547 [-] on Build GRCm38 | |

Ncam1
Genomic Map