siRNA Details
| Gene Symbol: 1700026J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI036213.1 | - | 49 | |||
| AK006389.1 | - | 8 | 997 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700026J12Rik| Gene Symbol: 1700026J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI036213.1 | - | 49 | |||
| AK006389.1 | - | 8 | 997 | |||