siRNA Details
| Gene Symbol: E030019B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045082.1 | 6 | 1626 | |||
| GenBank mRNA | AK053157.1 | - | 3 | 1901 | ||
| AK138262.1 | - | 93 | ||||
| BC064451.1 | - | 6 | 1466 | |||
| KF856953.1 | - | 7 | 1661 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E030019B13Rik| Gene Symbol: E030019B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045082.1 | 6 | 1626 | |||
| GenBank mRNA | AK053157.1 | - | 3 | 1901 | ||
| AK138262.1 | - | 93 | ||||
| BC064451.1 | - | 6 | 1466 | |||
| KF856953.1 | - | 7 | 1661 | |||