siRNA Details
| Gene Symbol: E030019B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045082.1 | 6, 7 | 1668 | |||
| GenBank mRNA | AK053157.1 | - | 4, 5 | 2079 | ||
| AK138262.1 | - | 2047 | ||||
| BC064451.1 | - | 6, 7 | 1508 | |||
| KF856953.1 | - | 7, 8 | 1703 | |||

E030019B13Rik
Genomic Map