siRNA Details
| Gene Symbol: Snapc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048571.1 | - | 1 | 1176 | ||
| AK053153.1 | - | 1 | 1860 | |||
| AK171985.1 | - | 1 | 1228 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snapc3| Gene Symbol: Snapc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048571.1 | - | 1 | 1176 | ||
| AK053153.1 | - | 1 | 1860 | |||
| AK171985.1 | - | 1 | 1228 | |||