siRNA Details
| Gene Symbol: Gm13264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140051.1 | - | 4 | 660 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13264| Gene Symbol: Gm13264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140051.1 | - | 4 | 660 | ||