siRNA Details
| Gene Symbol: Whsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050988.1 | - | 1 | 315 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV567816.1 | - | 20 | Chr. 5 - 33896328 - 33896853 [+] on Build GRCm38 | ||

Whsc1
Genomic Map