siRNA Details
| Gene Symbol: Gm10125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138232.1 | - | 2621 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10125| Gene Symbol: Gm10125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138232.1 | - | 2621 | |||