siRNA Details
| Gene Symbol: 9530041E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020593.1 | - | 1 | 481 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722105.1 | - | 1 | 481 | Chr. 7 - 27282184 - 27282889 [+] on Build GRCm38 | |

9530041E20Rik
Genomic Map