siRNA Details
| Gene Symbol: Gm33115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049687.1 | - | 3 | 726 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU936710.1 | - | 3 | 736 | Chr. 13 - 43843960 - 43846101 [+] on Build GRCm38 | |

Gm33115
Genomic Map