siRNA Details
| Gene Symbol: Peak1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK153735.1 | - | 2 | 374 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB638995.1 | - | 2 | 368 | Chr. 9 - 56257892 - 56267149 [+] on Build GRCm38 | |

Peak1os
Genomic Map