siRNA Details
| Gene Symbol: 1700003M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040647.1 | 2 | 260 | |||
| GenBank mRNA | AK005651.1 | - | 2 | 261 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705842.1 | - | 2 | 258 | Chr. 4 - 129960376 - 129963520 [+] on Build GRCm38 | |

1700003M07Rik
Genomic Map