siRNA Details
| Gene Symbol: 1700003M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040647.1 | 4 | 472 | |||
| NR_040648.1 | 3 | 272 | ||||
| GenBank mRNA | AA198428.1 | - | 3 | 283 | ||
| AK005651.1 | - | 4 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700003M07Rik| Gene Symbol: 1700003M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040647.1 | 4 | 472 | |||
| NR_040648.1 | 3 | 272 | ||||
| GenBank mRNA | AA198428.1 | - | 3 | 283 | ||
| AK005651.1 | - | 4 | 473 | |||